missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ GFI1 HEK293 Cell Transient Overexpression Lysate (Nondenatured)
Human overexpression lysate
Brand: Abnova™ L098T6.100ug
This item is not returnable.
View return policy
Specifications
HEK293 | |
GFI1 HEK293 Cell Transient Overexpression Lysate(Non-Denatured) | |
Modified RIPA Lysis Buffer:50 mM Tris-HCl pH 7.4, 150 mM NaCl, 1mM EDTA, 1% Triton X-100, 0.1% SDS, 1% Sodium deoxycholate, 1mM PMSF. ;Modified RIPA Lysis Buffer:50 mM Tris-HCl pH 7.4, 150 mM NaCl, 1mM EDTA, 1% Triton X-100, 0.1% SDS, 1% Sodium deoxycholate, 1mM PMSF. | |
pCMV-GFI1 full length | |
100 μg | |
Human | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. |
Immunoprecipitation, Western Blot | |
NM_005263 | |
Theoretical M.W. 45kDa | |
Transient overexpression cell lysate was tested with anti-GFI1 antibody (Mfr. No. H00002672-M01) by Western Blot. | |
In modified RIPA Lysis Buffer. | |
2 mg/mL |
Safety and Handling
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
Supplemental information
- 100UG GFI1 HEK293 Cell Transient Overexpression Ly sate(Non-Denatured)
Signal Word
- Warning
Hazard Category
- Aspiration toxicity Category 1
- Flammable liquid Category 3
Hazard Statement
- H226-Flammable liquid and vapour.
- H304-May be fatal if swallowed and enters airways.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P273-Avoid release to the environment.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/soap.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P312-Call a POISON CENTER/doctor/if you feel unwell.
- P332+P313-If skin irritation occurs: Get medical advice/attention.
- P337+P313-If eye irritation persists: Get medical advice/attention.
Supplemental information
- MIXTURE LIST-Contains : Dodecane