Learn More
Aalborg™ GFM Analog Mass Flowmeter
Designed to read flow rates of gases in a low cost and convenient manner. Aalborg™ GFM Analog Mass Flowmeter has rugged design coupled with instrumentation grade accuracy to provide versatile and economical means of flow measurement.
Brand:  Aalborg™ GFMS-010016
Product Code. 15284868
  • 1890.00 € / Each
Estimated Shipment: 20-03-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Rigid metallic construction
  • Built-in tiltable LCD readout
  • Circuit protection
  • Can be used as portable device
  • Engineering units or 0 to 100% displays
  • TIO - Totalizer option
Specifications
Specifications
Mass Flowmeter
0 to 200 LPM
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
View accessory products