Learn More
Thermo Scientific™ GFM Pro Gas Flowmeter
Measure and monitor the flow of all types of gases quickly and efficiently with the Thermo Scientific™ GFM Pro Gas Flowmeter.
Brand:  Thermo Scientific™ 66002-010
1141.66 EUR valid until 2025-12-31
Use promo code "25339" to get your promotional price.
Product Code. 10744186
  • 1155.00 € / Each
In Stock
to see stock.
Explore more special offers
Add to basket
Description
Description
Measure and monitor the flow of all types of laboratoty gases quickly and efficiently with the Thermo Scientific™ GFM Pro Gas Flowmeter. Specifically designed for use with GC systems, this CE-certified flowmeter has a measurement range of 0.5–500mL/min and measures the volumetric flow of any gas with an accuracy of +/- 2% or +/- 0.2mL/min, whichever is greater. Its easy-to-use interface features an over-range warning indicator and automatic shutoff, and its explosion-proof rating allows use with flammable and explosive gases. The compact ergonomic design enables use as a hand-held or benchtop unit and features side grips for added durability.
Additional Product Information
  • Data output via USB port
  • Calibration: traceable to NIST primary standards
  • Uses two AA batteries
  • Recalibration service available
TRUSTED_SUSTAINABILITY
Specifications
Specifications
±2% of Flow or ± 0.05 mL/min., whichever is greater
Electronic Gas Flowmeter
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Thermo Scientific™ GFM Pro Gas Flowmeter > Electronic gas flowmeter