missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Brady™ GHS Symbol - Acute Toxicity, Individual Markers on a Roll
Pre-printed labels to identify your chemical product containers.
Brand: Brady™ 834188
65.03 EUR valid until 2024-12-31
Use promo code "21676" to get your promotional price.
Description
- High-quality labels that can withstand the most demanding environments
- Highly resistant to bad weather, abrasion and chemical products
Specifications
PIC 1809-21*21-B7541-RLL-DC | |
Polyester | |
Permanent Acrylic | |
0.099 mm | |
-40°C to 110°C | |
Black on White |
Gloss | |
250/Pk. | |
21 mm | |
21 mm | |
Diamond |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction