Learn More
Brady™ GHS Symbol - Compressed Gas, Continuous roll
Pre-printed labels to identify your chemical product containers.
Brand:  Brady™ 814026
Product Code. 15654227
  • 441.00 € / Pack of 250
Estimated Shipment: 03-02-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • High-quality labels that can withstand the most demanding environments
  • Highly resistant to bad weather, abrasion and chemical products
Specifications
Specifications
PIC 1804-100*100-B7541-RLL-C
Polyester
Permanent Acrylic
0.099 mm
-40°C to 110°C
Black on White
Gloss
250/Pk.
100 mm
100 mm
Diamond