Learn More
Brady™ GHS Symbol - Hazardous to Aquatic Environment, Continuous Roll
Pre-printed labels to identify your chemical product containers.
Brand:  Brady™ 814028
399.06 EUR valid until 2024-12-20
Use promo code "21731" to get your promotional price.
Product Code. 15674227
  • 441.00 € / Pack of 250
In Stock
to see stock.
Explore more special offers
Add to basket
Description
Description
  • High-quality labels that can withstand the most demanding environments
  • Highly resistant to bad weather, abrasion and chemical products
Specifications
Specifications
Gloss
250/Pk.
100 mm
100 mm
Diamond
Polyester
Permanent Acrylic
0.099 mm
-40°C to 110°C
Black on White