Learn More
Fisherbrand™ PTFE Giant Stir Bar
Designed for use in large containers and vials. These giant PTFE-coated magnetic stir bares are autoclavable.
Brand:  Fisherbrand™ 001.257
 View more versions of this product
Product Code. 10470011
  • 13.60 € / Each
Estimated Shipment: 24-01-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Stable, general-purpose magnetic stir bar
  • Designed for use in large containers and vials
  • Autoclavable
Specifications
Specifications
Autoclavable
White
Containers & Vessels / Stirrer Hot Plates
57 mm
Cylinder
Certificate of Conformance, Certificate of Analysis, Product Data Sheet, Material Safety Data Sheet (MSDS)
PTFE encapsulated Alnico V magnet core
27 mm
1/Ea.