missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Giantin Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
145.00 € - 410.00 €
Specifications
Antigen | Giantin |
---|---|
Concentration | 0.54 mg/mL |
Applications | Immunofluorescence, Immunocytochemistry |
Classification | Polyclonal |
Conjugate | Unconjugated |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16831815
|
Proteintech
22270-1-AP-20UL |
20 μL |
145.00 €
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16821815
|
Proteintech
22270-1-AP-150UL |
150 μL |
410.00 €
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Golgin subfamily B member 1 (GOLGB1) is a single pass type I membrane protein located in the golgi apparatus membrane. GOLGB1 may participate in forming intercisternal cross-bridges of the Golgi complex.Specifications
Giantin | |
Immunofluorescence, Immunocytochemistry | |
Unconjugated | |
Rabbit | |
Human | |
Q14789 | |
2804 | |
Peptide | |
Primary | |
-20°C | |
GOLGB1 |
0.54 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
GCP, GCP372, giantin, GOLGB1, Golgin subfamily B member 1, GOLIM1, MacroGolgin | |
GOLGB1 | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title