missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ Advanced RPMI 1640 Medium
GREENER_CHOICE

Advanced RPMI (Roswell Park Memorial Institute) 1640 is a widely used basal medium that allows the culture of mammalian cells with reduced Fetal Bovine Serum (FBS) supplementation.

Brand:  Gibco™ 12633012

 View more versions of this product

Product Code. 11520446

  • 38.31 € / 500mL

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers

Requires : Supplementation
This item is not returnable. View return policy

Description

Description

Advanced RPMI (Roswell Park Memorial Institute) 1640 is a widely used basal medium that allows the culture of mammalian cells with reduced Fetal Bovine Serum (FBS) supplementation. Compared to classic RPMI 1640, serum supplementation can be reduced by 50-90% with no change in growth rate or morphology.i Cells successfully cultured in Advanced RPMI 1640, with no adaptation, include Sp2, Vero, Raji, Daudi, and Jurkat.

This Advanced RPMI 1640 is manufactured as follows:

With: Glucose, Non-essential Amino Acids, Sodium Pyruvate, Phenol Red

Without: L-glutamine, HEPES

The complete formulation is available.

Gibco™ Advanced RPMI 1640 is unique from other media due to addition of the following ingredients to allow for serum reduction: ethanolamine, glutathione, ascorbic acid, insulin, transferrin, AlbuMAX™ I lipid-rich bovine serum albumin for cell culture, and the trace elements sodium selenite, ammonium metavanadate, cupric sulfate, and manganous chloride.

Product Use

For Research Use Only. Not intended for animal or human diagnostic or therapeutic use.

Advanced RPMI 1640 requires supplementation with 1-5% Fetal Bovine Serum and 4 mM L-glutamine or GlutaMAX™ supplement. Many cell lines do not require adaptation to this media. The FBS concentration must be optimized for each cell line to obtain maximum serum reduction. Advanced RPMI 1640 uses a sodium bicarbonate buffer system (3.7 g/L) and therefore requires a 5-10% CO2 environment to maintain physiological pH.

iAdvanced Media: Reduced Sera Solutions. Quest, 2004. 1.3:26-30.

Order Info

Shipping Condition: Room Temperature

Compliance

Gibco Advanced RPMI 1640 is manufactured at a cGMP compliant facility located in Grand Island, New York. The facility is registered with the FDA as a medical device manufacturer and is certified to the ISO 13485 standards.

TRUSTED_SUSTAINABILITY
Specifications

Specifications

Storage conditions: 2-8° C. Protect from light
Shipping conditions: Ambient
Shelf life: 12 months from date of manufacture
Animal Origin
Liquid
Reduced Serum
Sterile-filtered
No Glutamine, No HEPES
Gibco™
Room Temperature
Sp2, Vero, Raji, Daudi, and Jurkat
1 X
RPMI 1640 Medium (Roswell Park Memorial Institute 1640 Medium)
Sterile-filtered
Glucose, Phenol Red, Sodium Pyruvate, Non Essential Amino Acids (NEAA)
cGMP-compliant under the ISO 13485 standard
500 mL
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only. Not for use in diagnostic procedures.