Learn More
Gibco™ Advanced RPMI 1640 Medium
Advanced RPMI (Roswell Park Memorial Institute) 1640 is a widely used basal medium that allows the culture of mammalian cells with reduced Fetal Bovine Serum (FBS) supplementation.
Brand: Gibco™ 12633020
309.67 EUR valid until 2025-02-21
Use promo code "24115" to get your promotional price.
Description
Advanced RPMI (Roswell Park Memorial Institute) 1640 is a widely used basal medium that allows the culture of mammalian cells with reduced Fetal Bovine Serum (FBS) supplementation. Compared to classic RPMI 1640, serum supplementation can be reduced by 50-90% with no change in growth rate or morphology.i Cells successfully cultured in Advanced RPMI 1640, with no adaptation, include Sp2, Vero, Raji, Daudi, and Jurkat.
This Advanced RPMI 1640 is manufactured as follows:
With: Glucose, Non-essential Amino Acids, Sodium Pyruvate, Phenol Red
Without: L-glutamine, HEPES
The complete formulation is available.
Gibco™ Advanced RPMI 1640 is unique from other media due to addition of the following ingredients to allow for serum reduction: ethanolamine, glutathione, ascorbic acid, insulin, transferrin, AlbuMAX™ I lipid-rich bovine serum albumin for cell culture, and the trace elements sodium selenite, ammonium metavanadate, cupric sulfate, and manganous chloride.
Product Use
For Research Use Only. Not intended for animal or human diagnostic or therapeutic use.
Advanced RPMI 1640 requires supplementation with 1-5% Fetal Bovine Serum and 4 mM L-glutamine or GlutaMAX™ supplement. Many cell lines do not require adaptation to this media. The FBS concentration must be optimized for each cell line to obtain maximum serum reduction. Advanced RPMI 1640 uses a sodium bicarbonate buffer system (3.7 g/L) and therefore requires a 5-10% CO2 environment to maintain physiological pH.
iAdvanced Media: Reduced Sera Solutions. Quest, 2004. 1.3:26-30.
Order Info
Shipping Condition: Room Temperature
Compliance
Gibco Advanced RPMI 1640 is manufactured at a cGMP compliant facility located in Grand Island, New York. The facility is registered with the FDA as a medical device manufacturer and is certified to the ISO 13485 standards.
Specifications
Storage conditions: 2-8° C. Protect from light Shipping conditions: Ambient Shelf life: 12 months from date of manufacture |
|
Animal Origin | |
Liquid | |
Reduced Serum | |
Sterile-filtered | |
No Glutamine, No HEPES | |
Gibco™ |
Sp2, Vero, Raji, Daudi, and Jurkat | |
1 X | |
RPMI 1640 Medium (Roswell Park Memorial Institute 1640 Medium) | |
Sterile-filtered | |
Glucose, Phenol Red, Sodium Pyruvate, Non Essential Amino Acids (NEAA) | |
cGMP-compliant under the ISO 13485 standard | |
10 x 500 mL |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only. Not for use in diagnostic procedures.