Learn More
Gibco™ Antibiotic-Antimycotic (100X)
Solution contains penicillin, streptomycin, and Gibco Amphotericin B to prevent cell culture contamination.
Brand: Gibco™ 15240062
86.69 EUR valid until 2025-02-21
Use promo code "24115" to get your promotional price.
Description
Gibco™ Antibiotic-Antimycotic is used to prevent bacterial and fungal contamination. This solution contains 10,000 units/mL of penicillin, 10,000 μg/mL of streptomycin, and 25 μg/mL of Gibco Amphotericin B. The antibiotics penicillin and streptomycin prevent bacterial contamination of cell cultures due to their effective combined action against gram-positive and gram-negative bacteria. Amphotericin B prevents fungal contamination of cell cultures due to its inhibition of multi-cellular fungus and yeast.
Penicillin was originally purified from the fungus Penicillium and acts by interfering directly with the turnover of the bacteria cell wall and indirectly by triggering the release of enzymes that further alter the cell wall. Streptomycin was originally purified from Streptomyces griseus. It acts by binding to the 30S subunit of the bacterial ribosome leading to inhibition of protein synthesis and death in susceptible bacteria. Amphotericin B is an antifungal agent that prevents the growth of fungi and yeast by causing an increase in fungal plasma membrane permeability.
Order Info
Shipping Condition: Dry Ice
Specifications
Storage conditions: -5°C to -20°C Shipping conditions: Dry ice Shelf life: 12 months from date of manufacture |
|
Mammalian Cell Culture | |
Antibiotic-Antimycotic | |
Sterile-filtered | |
Dry Ice |
100 X | |
Liquid | |
Sterile-filtered | |
100 mL |
Safety and Handling
- Antibiotic Antimycotic
Signal Word
- Danger
Hazard Category
- Reproductive toxicity Category 1
- Respiratory sensitiser Category 1
- Skin sensitiser Category 1
- Specific target organ toxicity Category 2
Hazard Statement
- H317-May cause an allergic skin reaction.
- H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
- H360-May damage fertility or the unborn child.
- H371-May cause damage to organs.
Precautionary Statement
- P201-Obtain special instructions before use.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P273-Avoid release to the environment.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/soap.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P308+P311-IF exposed or concerned: Call a POISON CENTER/doctor/.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
- P333+P313-If skin irritation or rash occurs: Get medical advice/attention.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
- MIXTURE LIST-Contains : penicillin G sodiumsalt, streptomycin sulphate
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only. Not for use in diagnostic procedures.