missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ Bovine Serum, heat inactivated, New Zealand origin
GREENER_CHOICE

Collected from cattle typically 12-24 months of age

35.98 € - 108.00 €

Specifications

Content And Storage Storage conditions: ≤-10°C
Shipping conditions: Frozen
Age 12 to 24 months
Species Cattle/Bovine
Country of Origin New Zealand
Form Liquid
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11580636
View Documents
Gibco™
26170035
100 mL
35.98 €
100mL
Please to purchase this item. Need a web account? Register with us today!
11590636
GREENER_CHOICE
View Documents
Gibco™
26170043
500 mL
108.00 €
500mL
Please to purchase this item. Need a web account? Register with us today!
Description

Description

  • Sera Category: Secure
  • Origin: New Zealand

Cell Culture, Mammalian Cell Culture

Specifications

Specifications

Storage conditions: ≤-10°C
Shipping conditions: Frozen
Cattle/Bovine
Liquid
Adult Bovine Serum
Sterile-filtered
Gibco™
12 to 24 months
New Zealand
Bottle
Heat-inactivated
Sterile-filtered
Frozen
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.