missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ CD Hybridoma AGT™ Medium

Animal origin-free, chemically defined, protein-free medium

Brand:  Gibco™ 12372025

142.20 EUR valid until 2025-04-18
Use promo code "24107" to get your promotional price.


 View more versions of this product

Product Code. 10710204

  • 158.00 € / 1 L

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description
Show More
Specifications

Specifications

Myeloma Cells, Hybridomas
Powder (AGT)
Serum-free
Sodium Pyruvate, Sodium Bicarbonate
AGT™, Gibco™
Wet Ice
Storage conditions: 2-8°C, dark and dry
Shipping conditions: Gel packs
Shelf life: 24 months from date of manufacture
Animal Origin-free, Chemically-defined, Protein-free
Hybridoma Medium
Non-sterile
No Glutamine, No Phenol Red
1 L
≤10 EU/mL, ≤ 10 EU/ml
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Manuals

Certificates

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number3083049 Certificate TypeCertificate of Analysis Date30/01/2025 Catalog Number12372003,12372025,12372001,12372017
Lot Number2387907 Certificate TypeCertificate of Analysis Date29/01/2025 Catalog Number12372003,12372025,12372001,12372017
Lot Number2481054 Certificate TypeCertificate of Analysis Date29/01/2025 Catalog Number12372003,12372025,12372001,12372017
Lot Number2519529 Certificate TypeCertificate of Analysis Date29/01/2025 Catalog Number12372003,12372025,12372001,12372017
Lot Number2531012 Certificate TypeCertificate of Analysis Date29/01/2025 Catalog Number12372003,12372025,12372001,12372017
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Gibco™ CD Hybridoma AGT™ Medium > 1L

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only. Not for use in diagnostic procedures.