Learn More
Gibco™ CD Hybridoma AGT™ Medium
Animal origin-free, chemically defined, protein-free medium
Brand: Gibco™ 12372001
Description
Gibco CD Hybridoma Medium is specifically developed for the ability to support the growth of hybridomas for antibody production.
- Ability to support suspension and stationary hybridomas and myelomas
- Formulated without L-glutamine for stability
- Animal origin-free, protein-free and chemically defined formulation
- Convenient, easy-to-use Advanced Granulation Technology (AGT™) format
Antibody and Cell Culture Production, Bioproduction, Cell Culture, Mammalian Cell Culture
Order Info
Shipping Condition: Wet ice

Specifications
Myeloma Cells, Hybridomas | |
1 X | |
Hybridoma Medium | |
Non-sterile | |
No Glutamine, No HEPES, No Phenol Red | |
AGT™, Gibco™ | |
Wet Ice | |
Calcium, Magnesium, Sodium Phosphate | |
Storage conditions: 2-8°C, dark and dry Shipping conditions: Gel packs Shelf life: 24 months from date of manufacture |
Animal Origin-free, Chemically-defined, Protein-free | |
Powder (AGT) | |
Serum-free | |
Sodium Pyruvate, Sodium Bicarbonate | |
3.66 g/L | |
100 L | |
Low | |
Certified | |
CO2 |
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
3083049 | Certificate of Analysis | 30/01/2025 | 12372003,12372025,12372001,12372017 |
2387907 | Certificate of Analysis | 29/01/2025 | 12372003,12372025,12372001,12372017 |
2481054 | Certificate of Analysis | 29/01/2025 | 12372003,12372025,12372001,12372017 |
2519529 | Certificate of Analysis | 29/01/2025 | 12372003,12372025,12372001,12372017 |
2531012 | Certificate of Analysis | 29/01/2025 | 12372003,12372025,12372001,12372017 |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only. Not for use in diagnostic procedures.