missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ DMEM, high glucose
GREENER_CHOICE

Código de producto. 15480564
Click to view available options
Quantity:
500 mL
1000 mL
5 L
10 L
Tamaño de la unidad:
1000mL
Each
Este artículo no se puede devolver. Vea la política de devoluciones

Código de producto. 15480564

Marca: Gibco™ 11965084

Please to purchase this item. Need a web account? Register with us today!

Edge
Este artículo no se puede devolver. Vea la política de devoluciones

Requires

Supplementation

DMEM (Dulbecco's Modified Eagle Medium) is a widely used basal medium for supporting the growth of many different mammalian cells.

DMEM (Dulbecco's Modified Eagle Medium) is a widely used basal medium for supporting the growth of many different mammalian cells. Cells successfully cultured in DMEM include primary fibroblasts, neurons, glial cells, HUVECs, and smooth muscle cells, as well as cell lines such as HeLa, 293, Cos-7, and PC-12. Life Technologies offers a variety of DMEM modifications for a range of cell culture applications. Find the right formulation using the media selector tool.

This DMEM is modified as follows:

With: High Glucose, L-glutamine, Phenol Red

Without: Sodium Pyruvate, HEPES

The complete formulation is available.

Using DMEM

DMEM is unique from other media as it contains 4 times the concentration of amino acids and vitamins than the original Eagle's Minimal Essential Medium. DMEM was originally formulated with low glucose (1 g/L) and sodium pyruvate, but is often used with higher glucose levels, with or without sodium pyruvate. DMEM contains no proteins, lipids, or growth factors. Therefore, DMEM requires supplementation, commonly with 10% Fetal Bovine Serum (FBS). DMEM uses a sodium bicarbonate buffer system (3.7 g/L), and therefore requires a 5–10% CO2 environment to maintain physiological pH.

Información de órdenes

Shipping Condition: Room Temperature

TRUSTED_SUSTAINABILITY

Especificaciones

Cell Line HeLa, 293, Cos-7, and PC-12
Cell Type Primary Fibroblasts, Neurons, Glial Cells, HUVECs, Smooth Muscle Cells
Classification Animal Origin-free
Concentration 1 X
Form Liquid
Product Type DMEM (Dulbecco's Modified Eagle Medium)
Serum Level Standard Serum Supplementation
Sterility Sterile-filtered
Sterilization Method Sterile-filtered
With Additives High Glucose, Glutamine, Phenol Red
Without Additives No HEPES, No Sodium Pyruvate
For Use With (Application) Mammalian Cell Culture
Glucose Concentration 4500 mg/L
Manufacturing Quality cGMP-compliant under the ISO 13485 standard
Product Line Gibco
Quantity 1000 mL
Shipping Condition Room Temperature
Content And Storage Storage conditions: 2-8° C. Protect from light
Shipping conditions: Ambient
Shelf life: 12 months from date of manufacture
Mostrar más Mostrar menos

For Research Use or Further Manufacturing. Not for diagnostic use or direct administration into humans or animals.

Corrección del contenido de un producto

Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.

Título del producto

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.

Gracias por ayudarnos a mejorar nuestra web. Su comentario ha sido enviado