missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ Donor Bovine Serum with Iron, New Zealand Origin
GREENER_CHOICE

Standard sera for robust cell lines

Brand:  Gibco™ 10371029

Product Code. 11503407

  • 145.00 € / 500mL

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

  • Sera Category: Secure Ferric citrate complex at 6.0mg/dL
  • Origin: New Zealand.
  • Endotoxin level: ≤10EU/mL
  • Collected from cattle typically 12-24 months of age

Cell Culture, Mammalian Cell Culture

Order Info

Shipping Condition: Dry Ice

TRUSTED_SUSTAINABILITY
Specifications

Specifications

Storage conditions: ≤-10°C
Shipping conditions: Frozen
12 to 24 months
Liquid
Fetal Bovine Serum
Standard (Sterile-filtered)
≤ 10 EU/mL
Frozen
Cattle/Bovine
New Zealand
Bottle
500 mL
Sterile
Gibco™
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

3 results found
Lot Number Certificate Type Date Catalog Number
Lot Number2509168 Certificate TypeCertificate of Analysis Date29/01/2025 Catalog Number10371029
Lot Number2897674 Certificate TypeCertificate of Analysis Date26/01/2025 Catalog Number10371029
Lot Number2977641 Certificate TypeCertificate of Analysis Date25/01/2025 Catalog Number10371029
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Gibco™ Donor Bovine Serum with Iron, New Zealand Origin > Quantity: 500 mL

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.