missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ DPBS, calcium, magnesium
GREENER_CHOICE

Dulbecco's phosphate-buffered saline (DPBS) is a balanced salt solution used for a variety of cell culture applications, such as washing cells before dissociation, transporting cells or tissue samples, diluting cells for counting, and preparing reagents.

31.31 € - 43.07 €

Specifications

Additive No Sodium Pyruvate
Classification Animal Origin-Free
Dilution 1 X
Product Line Gibco™
Product Type DPBS
View More Specs

Products 2
Product Code Brand Quantity Osmolality Price Quantity & Availability  
Product Code Brand Quantity Osmolality Price Quantity & Availability  
13492609
GREENER_CHOICE
View Documents
Gibco™
14040133
500 mL 270 to 300 mOsm/kg
31.31 €
Each
Please to purchase this item. Need a web account? Register with us today!
10217152
GREENER_CHOICE
View Documents
Gibco™
14040117
1000 mL 270 to 300 mOsm/kg
43.07 €
1 L
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Dulbecco's phosphate-buffered saline (DPBS) is a balanced salt solution used for a variety of cell culture applications, such as washing cells before dissociation, transporting cells or tissue samples, diluting cells for counting, and preparing reagents. Formulations with calcium and magnesium are generally used as transport media or for reagent preparation. We offer a variety of Gibco™ DPBS formulations for a range of cell culture applications.

This DPBS is modified as follows:

With: Calcium, Magnesium

Without: Phenol Red

Order Info

Shipping Condition: Room Temperature

Specifications

Specifications

No Sodium Pyruvate
1 X
DPBS
Washing Cells Before Dissociation, Transporting Cells or Tissue Samples, Diluting Cells for Counting, and Preparing Reagents
7.0 to 7.2
Sterile-filtered
cGMP-compliant under the ISO 13485 standard
Animal Origin-Free
Gibco™
Dulbecco's Phosphate Buffered Saline
Sustainable packaging
No Phenol Red
Liquid
Videos
SDS
Documents

Documents

Product Certifications

For Research Use or Further Manufacturing. Not for diagnostic use or direct administration into humans or animals.