Learn More
Gibco™ DPBS, no calcium, no magnesium
Dulbecco's Phosphate-Buffered Saline (DPBS) is a balanced salt solution used for a variety of cell culture applications, such as washing cells before dissociation, transporting cells or tissue, diluting cells for counting, and preparing reagents.
Brand: Gibco™ 14190359
280.68 EUR valid until 2024-09-28
Use promo code "21626" to get your promotional price.
Alert:
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21626” to get your promotional price
Description
Dulbecco's phosphate-buffered saline (DPBS) is a balanced salt solution used for a variety of cell culture applications, such as washing cells before dissociation, transporting cells or tissue samples, diluting cells for counting, and preparing reagents. Formulations without calcium and magnesium are required for rinsing chelators from the culture before cell dissociation.
This DPBS is modified as follows:
Without: Calcium, Magnesium, Phenol Red
Order Info
Shipping Condition: Room Temperature
Specifications
No Sodium Pyruvate | |
Storage conditions: 15°C to 30°C Shipping conditions: Ambient Shelf life: 12 months from date of manufacture |
|
Gibco™ | |
10 L | |
Mammalian Cell Culture | |
7.0 to 7.3 | |
Room Temperature | |
Liquid |
Animal Origin-Free | |
1 X | |
DPBS | |
Dulbecco's Phosphate Buffered Saline | |
270 - 300 mOsm/kg | |
No Phenol Red | |
Sterile-filtered | |
cGMP-compliant under the ISO 13485 standard |
For Research Use or Further Manufacturing. Not for diagnostic use or direct administration into humans or animals.