missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ Expi293™ Expression System Kit

Designed with transient expression technology for rapid and ultra high-yield protein production from mammalian cells. Gibco™ Expi293™ Expression System Kit is based on high density culture of Expi293F™ cells in Expi293™ Expression Medium.

Brand:  Gibco™ A14635

1601.40 EUR valid until 2024-11-30
Use promo code "21651" to get your promotional price.


Product Code. 13481059

  • 3140.00 € / Each

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

  • Rapid production of milligram to gram quantities of recombinant protein in mammalian cells
  • High density cultures of Expi293 cells that result in more expressing cells per milliliter of culture and higher protein yields
  • Native folding and mammalian post-translational modifications, such as glycosylation, of expressed proteins
  • Easy, robust culture and transfection protocols that readily fit into current transient expression workflows
  • Scalable expression that can be applied to cultures from<1mL to >10L, thus generating the amount of protein needed
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Expi293F™
Transfection
CMV
Mammalian
Protein Expression
33
Expression System Kit
1 kit
2 vials frozen Expi293F cells
1 bottle 1000 mL Expi293 Expression Medium
1 ExpiFectamine 293 transfection kit for 1 L of culture
1 vial Antibody Expressing Positive Control Vector
1 bottle 100 mL Opti-MEM Reduced Serum Medium

Store all components at +2°C to +8°C, except frozen cells are stored in liquid nitrogen
293-derived Cells
Lipid-based Transfection
Transient
Protein Production, Constitutive Expression
Expi293™
Untagged
Expi293™ Expression System
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

For Research Use Only. Not for use in diagnostic procedures.