Learn More
Gibco™ Fetal Bovine Serum, certified, heat inactivated, United States
Important Update: We've recently introduced Premium Plus Fetal Bovine Serum (FBS) specification-based products. These improved products are alternatives to our Certified US origin, New Zealand origin, and Qualified Australia origin SKUs. Product stability, performance, manufacturing, storage, and transport processes are unchanged.
Brand: Gibco™ A3840002
Description
Gibco fetal bovine sera offer excellent value for basic cell culture, specialty research, and specific assays, earning the trust of researchers with consistent quality and award-winning support that helps meet your research needs and budget requirements
- Sera Category: Premium (Performance Plus)
- Origin: United States
- Endotoxin level: ≤5 EU/mL
- Hemoglobin level: ≤15 mg/dL
Order Info
Shipping Condition: Dry Ice
Specifications
Storage conditions: ≤-10°C Shipping conditions: Frozen |
|
Fetal | |
United States | |
Fetal Bovine Serum | |
Heat-inactivated | |
Triple-filtered, 0.1 μm | |
≤15 mg/dL | |
One Shot™ |
Fetal Bovine Serum, heat inactivated, certified, One Shot™, United States | |
Bovine | |
One Shot | |
10 x 50 mL | |
Sterile | |
≤5 EU/mL | |
Certified | |
Frozen |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.