Learn More
Gibco™ Embryonic stem-cell FBS, qualified, USDA-approved regions
Embryonic stem-cell FBS, qualified, USDA-approved regions is US certified performance plus sera for broad range of cell types, especially sensitive cell lines.
734.00 €
Specifications
Content And Storage | Storage conditions: ≤-10°C Shipping conditions: Frozen |
---|---|
Age | Fetal |
Species | Cattle/Bovine |
Country of Origin | Mexico |
Form | Liquid |
Product Code | Brand | Quantity | Format | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Format | Price | Quantity & Availability | ||||
11518986
|
Gibco™
10439024 |
500 mL | Bottle | N/A | |||||
Description
Gibco™ fetal bovine sera offers excellent value for basic cell culture, specialty research, and specific assays, earning the trust of researchers with consistent quality and award-winning support that helps meet your research needs and budget requirements
- Sera Category: Specialty
- Origin: Countries meeting USDA importation requirements
- Endotoxin level: ≤50 EU/mL (levels routinely ≤10 EU/mL)
- Hemoglobin level: ≤25 mg/dL
Specially tested for the ability to sustain undifferentiated cellular morphology of embryonic stem cells.
Order Info
Shipping Condition: Dry Ice
Specifications
Storage conditions: ≤-10°C Shipping conditions: Frozen |
|
Cattle/Bovine | |
Liquid | |
Standard (Sterile-filtered) | |
<50 EU/mL | |
ES Cell-Qualified |
Fetal | |
Mexico | |
Fetal Bovine Serum | |
Sterile | |
≤25 mg/dL | |
Frozen |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.