missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Gibco™ Amphotericin B
Formulation of amphotericin B with sodium deoxycholate as an emulsifying agent to maintain the suspension
23.31 € - 43.19 €
Specifications
Concentration | 0.25 to 2.5 μg/mL |
---|---|
Form | Liquid |
Product Type | Antifungal |
For Use With (Application) | Prevention of Cell Culture Contamination |
Product Line | Fungizone™ |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11510496
|
Gibco™
15290018 |
20 mL |
23.31 €
20mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11520496
|
Gibco™
15290026 |
50 mL |
43.19 €
50mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
- Prevents growth of fungi by causing an increase in fungal plasma membrane permeability
- Gibco™ Fungizone™ Antimycotic is used to prevent contamination of cell cultures by yeast and multicellular fungi
- Contains 250μg of amphotericin B and 205μg of sodium deoxycholate per mL of distilled water
- Recommended working concentration ranges from 0.25 to 2.50μg/mL
Specifications
0.25 to 2.5 μg/mL | |
Antifungal | |
Fungizone™ |
Liquid | |
Prevention of Cell Culture Contamination | |
Dry Ice |
Safety and Handling
missing translation for 'shelfLife' : 12 Months
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
Supplemental information
- 4-Morpholinoacetophenone
Signal Word
- Warning
Hazard Category
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
Hazard Statement
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/soap.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P312-Call a POISON CENTER/doctor/if you feel unwell.
- P332+P313-If skin irritation occurs: Get medical advice/attention.
- P337+P313-If eye irritation persists: Get medical advice/attention.
Supplemental information
- MIXTURE LIST-Contains : 4-Morpholinoacetophenone
For Research Use Only. Not for use in diagnostic procedures.
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title