missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ Geneticin™ Selective Antibiotic (G418 Sulfate) (50 mg/mL)
GREENER_CHOICE

Product Code. 11558616
Click to view available options
Quantity:
20 mL
100 mL
Unit Size:
100mL
20mL
This item is not returnable. View return policy

Product Code. 11558616

Brand: Gibco™ 10131027

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Produced by bacterium Micromonospora rhodorangea and acts by binding ribosome, thus inhibiting protein synthesis in both prokaryotic and eukaryotic cells

Gibco Geneticin Selective Antibiotic (G418 Sulfate) is produced by the bacterium Micromonospora rhodorangea and acts by binding the ribosome, thus inhibiting protein synthesis in both prokaryotic and eukaryotic cells.

Resistance to Geneticin Selection Antibiotic (G418 Sulfate) is conferred by the E. coli APH(3)–I and APH(3)–II resistance genes.

  • Used as selective antibiotic in concentration range of 100 - 200μg/mL for bacteria, or 200 – 500μg/mL for most mammalian cells
  • Supplied as 50mg/mL solution in water

Cell Culture, Cloning, Drosophila S2 Cell Culture, High Five™ Cell Culture, Insect Cell Culture, Mammalian Cell Culture, Selection, Sf9 & Sf21 Cell Culture, Transfection, Transformation

Order Info

Shipping Condition: Room Temperature

Compliance

Manufactured at a cGMP compliant facility, which is registered with the FDA as a medical device manufacturer and is certified to ISO 13485 standards.

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Storage conditions: 2 to 8°C
Shipping conditions: Ambient
Shelf life: 24 months from date of manufacture
Cell Type Eukaryotic Cells, Prokaryotic Cells
Concentration 50 mg/mL
Culture Type Mammalian Cell Culture, Insect Cell Culture
Form Liquid
Product Type Antibiotic
Sterility Sterile-filtered
Sterilization Method Sterile-filtered
For Use With (Application) Eukaryotic Selection/Stable Cell Line Generation
Product Line Geneticin
Quantity 100 mL
Shipping Condition Room Temperature
Show More Show Less
compliance-icons
Product Identifier
  • GENETICIN
Signal Word
  • Danger
Hazard Category
  • Respiratory sensitiser Category 1
  • Skin sensitiser Category 1
Hazard Statement
  • H317-May cause an allergic skin reaction.
  • H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P272-Contaminated work clothing should not be allowed out of the workplace.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P284-¢In case of inadequate ventilation! wear respiratory protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P333+P313-If skin irritation or rash occurs: Get medical advice/attention.
  • P342+P311-If experiencing respiratory symptoms: Call a POISON CENTER/doctor/ .
  • P362+P364-Take off contaminated clothing and wash it before reuse.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contains : D-Streptamine

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.