missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ Geneticin™ Selective Antibiotic (G418 Sulfate), Powder

Produced by bacterium Micromonospora rhodorangea and acts by binding the ribosome, thus inhibiting protein synthesis in both prokaryotic and eukaryotic cells

160.00 € - 2160.00 €

Specifications

Content And Storage Storage conditions: 15 to 30°C
Shipping conditions: Ambient
Shelf life: 24 months from date of manufacture
Cell Type Eukaryotic Cells, Prokaryotic Cells
Concentration 10 to 50 μg/mL
Form Powder
Product Type Antibiotic
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
10463982
View Documents
Gibco™
11811023
1 g
160.00 €
1g
Please to purchase this item. Need a web account? Register with us today!
11528676
View Documents
Gibco™
11811031
5 g
508.00 €
5g
Please to purchase this item. Need a web account? Register with us today!
11548676
View Documents
Gibco™
11811098
25 g
2160.00 €
25g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Resistance to Geneticin™ Selective Antibiotic (G418 Sulfate) is conferred by the E. coli APH (3) – I and APH (3) – II resistance genes.

  • Used as selective antibiotic in concentration range of 100 - 200μg/mL for bacteria, or 200 – 500μg/mL for most mammalian cells
  • Supplied as powder and should be made into a stock solution of 10 – 50mg/mL in water

Cell Culture, Cloning, Drosophila S2 Cell Culture, High Five™ Cell Culture, Insect Cell Culture, Mammalian Cell Culture, Selection, Sf9 & Sf21 Cell Culture, Transfection, Transformation

Order Info

Shipping Condition: Room Temperature

Compliance

Manufactured at a cGMP compliant facility, which is registered with the FDA as a medical device manufacturer and is certified to ISO 13485 standards.

Specifications

Specifications

Storage conditions: 15 to 30°C
Shipping conditions: Ambient
Shelf life: 24 months from date of manufacture
10 to 50 μg/mL
Antibiotic
Geneticin™
Eukaryotic Cells, Prokaryotic Cells
Powder
Eukaryotic Selection⁄Stable Cell Line Generation
Room Temperature
Videos
Safety and Handling

Safety and Handling

missing translation for 'shelfLife' : 24 Months

Product Identifier
  • GENETICIN

Signal Word
  • Danger

Hazard Category
  • Respiratory sensitiser Category 1
  • Skin sensitiser Category 1

Hazard Statement
  • H317-May cause an allergic skin reaction.
  • H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.

Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P272-Contaminated work clothing should not be allowed out of the workplace.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P284-¢In case of inadequate ventilation! wear respiratory protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P333+P313-If skin irritation or rash occurs: Get medical advice/attention.
  • P342+P311-If experiencing respiratory symptoms: Call a POISON CENTER/doctor/ .
  • P362+P364-Take off contaminated clothing and wash it before reuse.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.

Supplemental information
  • MIXTURE LIST-Contains : D-Streptamine, O-2-amino-2,7-dideoxy-D-glycero-.alp ha.-D-gluco-heptopyranosyl-(1.fwdarw. 4)-O-[3-de

SDS
Documents

Documents

Product Certifications

For Research Use Only. Not for use in diagnostic procedures.