missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ Blasticidin S HCl (10 mg/mL)

Selective antibiotic in concentration range of 50G–100μg/mL

644.00 € - 1238.00 €

Specifications

Content And Storage Storage conditions: -5°C to -20°C (protect from light)
Shipping conditions: Dry ice
Shelf life: 9 months from date of manufacture
Cell Type Eukaryotic Cells, Prokaryotic Cells
Concentration 10 mg/mL
Culture Type Mammalian Cell Culture, Insect Cell Culture
Form Liquid
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
12172530
View Documents
Gibco™
A1113903
10 x 1 mL
644.00 €
10mL
Please to purchase this item. Need a web account? Register with us today!
10166793
View Documents
Gibco™
A1113902
20 mL
1238.00 €
Each
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Blasticidin S is a nucleoside antibiotic isolated from Streptomyces griseochromogenes. It is a potent inhibitor of protein synthesis in both prokaryotic and eukaryotic cells. Blasticidin S acts quickly by inhibiting peptide bond formation by the ribosome.

  • Used for prevention of contamination of cell cultures
  • Recommended working concentration ranges 1–20μg/mL depending on cell line
  • Provided as solution at 10mg/mL in 20 mM HEPES (pH 7.2–7.5)
  • Supplied in 10 vials each containing 1mL.
  • Resistance is conferred by expression of either one of two Blasticidin S deaminase genes: BSD or bsr

Cell Culture, Cloning, Drosophila S2 Cell Culture, Insect Cell Culture, Mammalian Cell Culture, Selection, Sf9 and Sf21 Cell Culture, Transfection, Transformation

Order Info

Shipping Condition: Dry Ice

Specifications

Specifications

Storage conditions: -5°C to -20°C (protect from light)
Shipping conditions: Dry ice
Shelf life: 9 months from date of manufacture
10 mg/mL
Liquid
Sterile-filtered
Gibco™
Eukaryotic Cells, Prokaryotic Cells
Mammalian Cell Culture, Insect Cell Culture
Antibiotic
HEPES
Videos
Safety and Handling

Safety and Handling

missing translation for 'shelfLife' : 9 Months

Product Identifier
  • Blasticidin S HCL

Signal Word
  • Warning

Hazard Category
  • Acute toxicity Category 4

Hazard Statement
  • H302-Harmful if swallowed.

Precautionary Statement
  • P264-Wash hands thoroughly after handling.
  • P270-Do not eat, drink or smoke when using this product.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P330-Rinse mouth.

Supplemental information
  • MIXTURE LIST-Contains : Blasticidin S HCL

SDS
Documents

Documents

Product Certifications

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.