missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ Grace's Insect Medium, supplemented
GREENER_CHOICE

Widely used for the growth of Spodeptera frugiperda cells, Sf9 and Sf21

Brand:  Gibco™ 11605094

93.50 EUR valid until 2024-12-31
Use promo code "21651" to get your promotional price.


 View more versions of this product

Product Code. 13426146

  • 110.00 € / 500mL

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers

Requires : Grace's Unsupplemented Insect Medium requires ambient CO2 in order to maintain physiological pH.
This item is not returnable. View return policy

Description

Description

Originally designed for the growth of cells from the Australian Emperor Gum Moth, Antherea eucalypti. Sf9 and Sf21 cells are routinely used to express recombinant protein using the Baculovirus Expression Vector System (BEVS).

  • Contains lactalbumin hydrolysate, yeastolate, L-glutamine, and sodium bicarbonate
  • Dual site cGMP manufacturing and quality system
  • Commonly supplemented with 10% Fetal Bovine Serum (FBS) before use

Cell Culture, Insect Cell Culture, Sf9 and Sf21 Cell Culture

Order Info

Shipping Condition: Room temperature

TRUSTED_SUSTAINABILITY
Specifications

Specifications

Storage conditions: 2°C to 8°C. Protect from light
Shipping conditions: Ambient
Shelf life: 12 months from date of manufacture
Insect Cell
Grace's Insect Medium
No Insect Hemolymph
Gibco™
Room Temperature
Sf21, Sf9
Liquid
Glutamine, Yeastolate, Lactalbumin Hydrolysate
Calcium, Magnesium
500 mL
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only. Not for use in diagnostic procedures.