Learn More
Gibco™ Ham's F-10 Nutrient Mix
Ham's F-10 Nutrient Mixture (F-10) was designed for serum-free growth of Chinese Hamster Ovary (CHO) cells.
Brand: Gibco™ 11550043
19.96 EUR valid until 2024-09-28
Use promo code "21626" to get your promotional price.
Alert:
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21626” to get your promotional price
Requires : Supplementation
This item is not returnable. View return policy
Description
Ham's F-10 Nutrient Mixture (F-10) was designed for serum-free growth of Chinese Hamster Ovary (CHO) cells. F-10 has since been used for serum-free growth of CHO cultures as well as serum-supplemented growth of other mammalian cells, including COS-7, primary rat astrocytes, and rat prostate epithelial cells.
This Ham's F-10 is modified as follows:
With: Phenol Red, L-glutamine
Without: HEPES
The complete formulation is available.
Using Ham's F-10
Compared to other basal media, F-10 contains a wider variety of components, including zinc, hypoxanthine, and thymidine. Serum-free growth of CHO cells in F-10 has led to a variety of improved formulations, such as Ham's F-12 Nutrient Mixture (F-12). F-10 contains no proteins or growth factors. Therefore, F-10 requires supplementation, commonly with 10% Fetal Bovine Serum (FBS). F-10 uses a sodium bicarbonate buffer system (1.2 g/L), and therefore requires a 5–10% CO2 environment to maintain physiological pH.
Order Info
Shipping Condition: Room Temperature
Specifications
Storage conditions: 2°C to 8°C (protect from light) Shipping conditions: Ambient Shelf life: 12 months from date of manufacture |
|
Primary Rat Astrocytes | |
1 X | |
Ham's F-10 Nutrient Mixture | |
High Glucose, Glutamine, Phenol Red, Sodium Pyruvate | |
Sustainable packaging | |
Gibco™ | |
Room Temperature |
CHO, COS-7, and rat prostate epithelial cells | |
Animal Origin-free | |
Liquid | |
Sterile-filtered | |
No HEPES | |
cGMP-compliant under the ISO 13485 standard | |
500 mL |
For Research Use Only. Not for use in diagnostic procedures.