missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ Ham's F-12 Nutrient Mix, powder

Product Code. p-4919907
Click to view available options
Quantity:
50 L
10 x 1 L
Unit Size:
Each
Pack of 10
This item is not returnable. View return policy

Product Code. 15480604

Brand: Gibco™ 21700075

Please to purchase this item. Need a web account? Register with us today!

Edge
This item is not returnable. View return policy

Ham's F-12 Nutrient Mixture (F-12) was designed for serum-free single-cell plating of Chinese Hamster Ovary (CHO) cells.

Ham's F-12 Nutrient Mixture (F-12) was designed for serum-free single-cell plating of Chinese Hamster Ovary (CHO) cells. F-12 has since been used for serum-free growth of CHO cultures as well as serum-supplemented growth of other mammalian cells, including chondrocytes and rat prostate epithelial cells. We offer a variety of Gibco™ F-12 modifications for a range of cell culture applications. Find the right formulation using the media selector tool.

This Ham's F-12 is modified as follows:

With: L-glutamine, Phenol Red

Without: Sodium Bicarbonate

The complete formulation is available.

Compared to other basal media, F-12 contains a wider variety of components, including zinc, putrescine, hypoxanthine, and thymidine. Serum-free growth of CHO cells in F-12 has led to a variety of improved formulations.

Gibco™ F-12 contains no proteins or growth factors. Therefore, F-12 requires supplementation, commonly with 10% Fetal Bovine Serum (FBS). F-12 uses a sodium bicarbonate buffer system (1.176 g/L) and therefore requires a 5-10% CO2 environment to maintain physiological pH. Powder forms of Gibco™ cell culture medium require sodium bicarbonate supplementation, pH adjustment, and filtration at the time of preparation (see protocol for details).

Please note that the shelf life on delivered products varies. This product has a minimum shelf life of 6 months at time of shipment. For more information regarding product shelf life, including details on specific product lots, please contact Customer Service.

Order Info

Shipping Condition: Room Temperature

Compliance

Gibco F-12 is manufactured at a cGMP compliant facility located in Grand Island, New York. The facility is registered with the FDA as a medical device manufacturer and is certified to ISO 13485 standards.

TRUSTED_SUSTAINABILITY

Specifications

Cell Type Primary Rat Astrocytes
Classification Animal Origin-free
Form Powder
Product Type Ham's F-12 Nutrient Mixture
Sterility Sterile-filtered
With Additives Glutamine, Phenol Red, Sodium Pyruvate
Without Additives No HEPES, No Sodium Bicarbonate
Manufacturing Quality cGMP-compliant under the ISO 13485 standard
Product Line Gibco
Quantity 10 x 1 L
Shipping Condition Room Temperature
Content And Storage Storage conditions: 2-8° C
Shipping conditions: Ambient
Shelf life: 36 months from date of manufacture
Show More Show Less

Not for human or animal therapeutic use. Uses other than the intended use may be a violation of local law. For in vitro diagnostic use.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.