Learn More
Gibco™ Ham's F-12K (Kaighn's) Medium
![GREENER_CHOICE](/content/dam/fishersci/glyphs/green_glyph.png)
Ham's F-12K (Kaighn's) Medium is a modification of Ham's F-12 Nutrient Mixture. Ham's F-12K (Kaighn's) Medium was developed for primary human hepatocytes, as well as for some rat and chicken liver cells in a reduced serum environment.
50.50 € - 454.00 €
Specifications
Content And Storage | Storage conditions: 2-8°C. Protect from light Shipping conditions: Ambient |
---|---|
Cell Type | Human Hepatocytes |
Classification | Animal Origin-free |
Concentration | 1 X |
Form | Liquid |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11580556
![]() |
Gibco™
21127022 |
500 mL |
50.50 €
500mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
10667440
![]() |
Gibco™
21127030 |
10 x 500 mL |
454.00 €
Pack of 10 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Ham's F-12K (Kaighn's) Medium is a modification of Ham's F-12 Nutrient Mixture. Ham's F-12K (Kaighn's) Medium was developed for primary human hepatocytes, as well as for some rat and chicken liver cells in a reduced serum environment.
This F-12K (Kaighn's) Medium is manufactured as follows:
With: L-glutamine, Phenol Red
Without: HEPES
The complete formulation is available.
Using Ham's F-12K
Ham's F-12K (Kaighn's) Medium contains many components not found in traditional basal media, such as putrescine, thymidine, hypoxanthine, zinc, and higher levels of all amino acids and sodium pyruvate. These additions allow the medium to be supplemented with very low levels of serum or defined components, for some cell types. Ham's F-12K (Kaighn's) Medium contains no proteins or growth factors, and is therefore often supplemented with growth factors and Fetal Bovine Serum (FBS). The FBS concentration must be optimized for each cell line. Ham's F-12K (Kaighn's) Medium uses a sodium bicarbonate buffer system (2.5 g/L), and therefore requires a 5–10% CO2 environment to maintain physiological pH.
Order Info
Shipping Condition: Room Temperature
Specifications
Storage conditions: 2-8°C. Protect from light Shipping conditions: Ambient |
|
Animal Origin-free | |
Liquid | |
Sterile-filtered | |
Low Glucose, Glutamine, Phenol Red, Sodium Pyruvate | |
cGMP-compliant under the ISO 13485 standard | |
Room Temperature |
Human Hepatocytes | |
1 X | |
Ham's F-12K (Kaighn's) Medium | |
Sterile-filtered | |
No HEPES | |
Gibco™ |
Safety and Handling
missing translation for 'shelfLife' : 12 Months From Date of Manufacture
- Silver chloride
Signal Word
- Warning
Hazard Category
- LONG-TERM AQUATIC HAZARD Chronic 1
- ACUTE AQUATIC Acute 1
- Substances and mixtures corrosive to metals Category 1
Hazard Statement
- H290-May be corrosive to metals.
- H400-Very toxic to aquatic life.
- H410-Very toxic to aquatic life with long lasting effects.
Precautionary Statement
- P273-Avoid release to the environment.
- P390-Absorb spillage to prevent material damage.
- P391-Collect spillage.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
- MIXTURE LIST-Contains : Silver chloride
For Research Use Only. Not for use in diagnostic procedures.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.