missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ HBSS, no calcium, no magnesium
GREENER_CHOICE

Hanks' Balanced Salt Solution (HBSS) is used for a variety of cell culture applications, such as washing cells before dissociation, transporting cells or tissue samples, diluting cells for counting, and preparing reagents.

39.27 €

Specifications

Additive No Sodium Pyruvate
Classification Animal Origin-Free
Product Line Gibco™
Product Type Hanks' Balanced Salt Solution (HBSS)
Solution Type Hank's Balanced Salt
View More Specs

Products 1
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
12082739
GREENER_CHOICE
View Documents
Gibco™
14170112
500 mL
39.27 €
Each
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Hanks' Balanced Salt Solution (HBSS) is used for a variety of cell culture applications, such as washing cells before dissociation, transporting cells or tissue samples, diluting cells for counting, and preparing reagents. Formulations without calcium and magnesium are required for rinsing chelators from the culture before cell dissociation.

We offer a variety of Gibco™ HBSS formulations for a range of cell culture applications.

This HBSS is modified as follows:

With: Phenol Red, Glucose

Without: Calcium, Magnesium

The complete formulation is available.

Order Info

Shipping Condition: Room Temperature

Specifications

Specifications

No Sodium Pyruvate
Gibco™
Hank's Balanced Salt
6.7 to 7.8
Room Temperature
Liquid
Animal Origin-Free
Hanks' Balanced Salt Solution (HBSS)
Mammalian Cell Culture
Phenol Red
Sterile-filtered
cGMP-compliant under the ISO 13485 standard
Videos
SDS
Documents

Documents

Product Certifications

For Research Use or Further Manufacturing. Not for diagnostic use or direct administration into humans or animals.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.