Learn More
Gibco™ HBSS, no calcium, no magnesium, no phenol red
Hanks' Balanced Salt Solution (HBSS) is used for a variety of cell culture applications, such as washing cells before dissociation, transporting cells or tissue samples, diluting cells for counting, and preparing reagents.
37.97 €
Specifications
Additive | No Sodium Pyruvate |
---|---|
Color | Clear |
Dilution | 1 X |
Product Line | Gibco™ |
Product Type | Hanks' Balanced Salt Solution (HBSS) |
Description
Hanks' Balanced Salt Solution (HBSS) is used for a variety of cell culture applications, such as washing cells before dissociation, transporting cells or tissue samples, diluting cells for counting, and preparing reagents. Formulations without calcium and magnesium are required for rinsing chelators from the culture before cell dissociation.
We offer a variety of Gibco™ HBSS formulations for a range of cell culture applications.
This HBSS is modified as follows:
With: Glucose
Without: Calcium, Magnesium, Phenol Red
The complete formulation is available.
Order Info
Shipping Condition: Room Temperature
Specifications
No Sodium Pyruvate | |
1 X | |
Hanks' Balanced Salt Solution (HBSS) | |
Mammalian Cell Culture | |
270 - 295 mOsm/kg | |
No Phenol Red | |
Liquid |
Clear | |
Gibco™ | |
Hank's Balanced Salt | |
Sustainable packaging | |
6.7 - 7.8 | |
Sterile-filtered | |
cGMP-compliant under the ISO 13485 standard |
For Research Use or Further Manufacturing. Not for diagnostic use or direct administration into humans or animals.