missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ Horse Serum, heat inactivated, New Zealand origin

EIA tested (negative) serum from a donor herd

29.62 € - 135.00 €

Specifications

Content And Storage Storage conditions: ≤-10°C, Shipping conditions: Frozen
Species Horse/Equine, Horse
Country of Origin New Zealand
Form Liquid
Product Type Horse Serum
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11540636
View Documents
Gibco™
26050070
100 mL
29.62 €
100mL
Please to purchase this item. Need a web account? Register with us today!
10368902
View Documents
Gibco™
26050088
500 mL
135.00 €
500mL
Please to purchase this item. Need a web account? Register with us today!
Description

Description

  • Gibco sera offers excellent value for basic cell culture, specialty research, and specific assays
  • It has earned the trust of researchers with consistent quality and award-winning support
  • Helps meet research needs and budget requirements
  • EIA tested (negative) serum from a donor herd
  • Shipped frozen, store at <10°C
  • Sera Category: Specialty
  • Origin: New Zealand

Order Info

Shipping Condition: Dry ice

Specifications

Specifications

Storage conditions: ≤-10°C, Shipping conditions: Frozen
New Zealand
Horse Serum
Gibco™
Horse/Equine, Horse
Liquid
Heat-inactivated
Frozen
Videos
SDS
Documents

Documents

Product Certifications

For Research Use or Further Manufacturing. Not for diagnostic use or direct administration into humans or animals.