Learn More
Gibco™ IMDM, GlutaMAX™ Supplement
IMDM (Iscove's Modified Dulbecco's Medium) is well suited for rapidly proliferating, high-density cell cultures, including Jurkat, COS-7, and macrophage cells.
Brand: Gibco™ 31980030
54.57 EUR valid until 2024-10-31
Use promo code "21744" to get your promotional price.
Description
IMDM (Iscove's Modified Dulbecco's Medium) is well suited for rapidly proliferating, high-density cell cultures, including Jurkat, COS-7, and macrophage cells. We offer a variety of IMDM modifications for a range of cell culture applications. Find the right formulation using the media selector tool.
This IMDM is modified as follows:
With: GlutaMAX™, Phenol Red
Without: α-thioglycerol, 2-mercaptoethanol
The complete formulation is available.
Using IMDM
IMDM, a modification of Dulbecco's Modified Eagle Medium, includes selenium as well as additional amino acids and vitamins. In addition, this unique medium lacks iron, with potassium nitrate replacing ferric nitrate. IMDM with GlutaMAX™ supplement minimizes toxic ammonia build-up and improves cell viability and growth in an easy-to-use format. IMDM contains no proteins, lipids, or growth factors. Therefore, IMDM requires supplementation, commonly with 10% Fetal Bovine Serum (FBS). IMDM uses a sodium bicarbonate buffer system (3.024 g/L), and therefore requires a 5–10% CO2 environment to maintain physiological pH.
Order Info
Shipping Condition: Room Temperature
Specifications
Storage conditions: 2-8° C. Protect from light Shipping conditions: Ambient Shelf life: 12 months from date of manufacture |
|
Animal Origin-free | |
Liquid | |
Sterile-filtered | |
No α-Thioglycerol, No 2-Mercaptoethanol | |
cGMP-compliant under the ISO 13485 standard | |
500 mL |
Jurkat, COS-7, and macrophage cells | |
1 X | |
IMDM (Iscove's Modified Dulbecco's Medium) | |
High Glucose, GlutaMAX, HEPES, Phenol Red, Sodium Pyruvate | |
Sustainable packaging | |
Gibco™, GlutaMAX™ | |
Room Temperature |
In vitro diagnostic