Learn More
Gibco™ IMDM, no phenol red
Iscove's Modified Dulbecco's Medium (IMDM) is well suited for rapidly proliferating, high-density cell cultures, including Jurkat, COS-7, and macrophage cells.
Brand: Gibco™ 21056023
16.48 EUR valid until 2024-09-28
Use promo code "21626" to get your promotional price.
Alert:
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21626” to get your promotional price
Requires : Supplementation
This item is not returnable. View return policy
Description
Iscove's Modified Dulbecco's Medium (IMDM) is well suited for rapidly proliferating, high-density cell cultures, including Jurkat, COS-7, and macrophage cells. We offer a variety of Gibco™ IMDM modifications for a range of cell culture applications. Find the right formulation using the media selector tool.
This IMDM is modified as follows:
With: L-glutamine
Without: Phenol Red, α-thioglycerol, 2-mercaptoethanol
The complete formulation is available.
IMDM, a modification of Dulbecco's Modified Eagle Medium, includes selenium as well as additional amino acids and vitamins. In addition, this unique medium lacks iron, with potassium nitrate replacing ferric nitrate.
Product Use
For Research Use Only: Not intended for animal or human diagnostic or therapeutic use.
IMDM contains no proteins, lipids, or growth factors. Therefore, IMDM requires supplementation, commonly with 10% Fetal Bovine Serum (FBS). IMDM uses a sodium bicarbonate buffer system (3.024 g/L) and therefore requires a 5-10% CO2 environment to maintain physiological pH.
Order Info
Shipping Condition: Room Temperature
Compliance
Gibco IMDM is manufactured at a cGMP compliant facility located in Grand Island, New York. The facility is registered with the FDA as a medical device manufacturer and is certified to ISO 13485 standards.
Specifications
Storage conditions: 2-8° C. Protect from light Shipping conditions: Ambient Shelf life: 12 months from date of manufacture |
|
Animal Origin-free | |
Liquid | |
Sterile-filtered | |
No Phenol Red, No α-Thioglycerol, No 2-Mercaptoethanol | |
cGMP-compliant under the ISO 13485 standard | |
500 mL |
Jurkat, COS-7, and macrophage cells | |
1 X | |
IMDM (Iscove's Modified Dulbecco's Medium) | |
High Glucose, Glutamine, HEPES, Sodium Pyruvate | |
Sustainable packaging | |
Gibco™ | |
Room Temperature |
For Research Use Only. Not for use in diagnostic procedures.