missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ IMDM, powder

Iscove's Modified Dulbecco's Medium (IMDM) is well suited for rapidly proliferating, high-density cell cultures, including Jurkat, COS-7, and macrophage cells.

Brand:  Gibco™ 12200069

 View more versions of this product

Product Code. 17102949

  • 146.00 € / 10 L

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers

Requires : Supplementation
This item is not returnable. View return policy

Description

Description

Iscove's Modified Dulbecco's Medium (IMDM) is well suited for rapidly proliferating, high-density cell cultures, including Jurkat, COS-7, and macrophage cells. We offer a variety of Gibco™ IMDM modifications for a range of cell culture applications. Find the right formulation using the media selector tool.

This IMDM is modified as follows:
With: L-glutamine, Phenol Red
Without: α-thioglycerol, 2-mercaptoethanol

The complete formulation is available.

IMDM, a modification of Dulbecco's Modified Eagle Medium, includes selenium as well as additional amino acids and vitamins. In addition, this unique medium lacks iron, with potassium nitrate replacing ferric nitrate.

Order Info

Shipping Condition: Room Temperature

TRUSTED_SUSTAINABILITY
Specifications

Specifications

Storage conditions: 2-8° C
Shipping conditions: Ambient
Shelf life: 36 months from date of manufacture
Animal Origin-free
IMDM (Iscove's Modified Dulbecco's Medium)
High Glucose, Glutamine, HEPES, Phenol Red, Sodium Pyruvate
cGMP-compliant under the ISO 13485 standard
10 L
Jurkat, COS-7, and macrophage cells
Powder
Sterile-filtered
No α-Thioglycerol, No 2-Mercaptoethanol
Gibco™
Room Temperature
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Not for human or animal therapeutic use. Uses other than the intended use may be a violation of local law.