missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Gibco™ Lab Armor™ Beads

Small, non-uniform metal beads comprised of dry, metallic thermal alloy for use in water baths designed to replace water in a water bath or ice in an ice bucket.
Brand: Gibco™ A1254301
258.00 EUR valid until 2025-12-31
Use promo code "24071" to get your promotional price.
This item is not returnable.
View return policy
Description
- Maintains uniform temperature better than water by transferring heat more slowly
- Can remain "always-on" and ready to use
- Bottles and tubes stay in place without need for racks, weights and floats
- Removes need for water bath maintenance such as emptying, cleaning and refilling bath
- Eliminates risk of contamination from standing water in water bath
- No condensation seen around caps and incubating bottles removed from dry bath
- Environmentally responsible alternative to water
- More energy efficient than water
- Draws less energy when run at 37 and 65°C
- Made from recyclable material that can be cleaned with ethanol, eliminating need for water bath germicides that can harm the environment
- Store at 15 to 30°C (shipped under ambient conditions)

Specifications
Reflective, Inert, Smooth | |
4 L | |
Bead | |
Gibco™ | |
Room Temperature |
Storage conditions: 15 – 30°C Shipping conditions: Ambient |
|
-80°C to +180°C | |
Energy efficient | |
Volume: 4 L | |
5 mm poly-disperse |
Safety and Handling
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
Supplemental information
- 4-Nitrobenzoyl chloride
Signal Word
- Danger
Hazard Category
- Serious eye damage/eye irritation Category 1
- Skin corrosion/irritation Category 1 B
Hazard Statement
- H314-Causes severe skin burns and eye damage.
- H318-Causes serious eye damage.
- EUH029-Contact with water liberates toxic gas.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
- P402+P404-Store in a dry place. Store in a closed container.
Supplemental information
- MIXTURE LIST-Contains : 4-Nitrobenzoyl chloride
Lab Armor™ Beads - Green Fact Sheet
File size: 140.3 kB
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Gibco™ Lab Armor™ Beads > 4L
For Research Use Only. Not for use in diagnostic procedures.
Spot an opportunity for improvement?Share a Content Correction