Learn More
Gibco™ LV-MAX™ Production Medium
Brand: Gibco™ A3583401
285.30 EUR valid until 2025-06-30
Use promo code "24074" to get your promotional price.
Description
Gibco LV-MAX Production Medium is a chemically defined, serum-free, protein-free medium, specially developed for high-density growth and transfection of Gibco Viral Production Cells (suspension-adapted HEK 293-derived cells) promoting high-titer lentiviral production.
- Supports growth of suspension Viral Production Cells to densities of over 1.0 x 107 cells/mL
- Enables sustained, high-level production of high-density transiently transfected cultures, achieving titer of up to 1 x 108 TU/mL (unconcentrated LVVs-GFP)
- Supports growth and transfection of Viral Production Cells in culture formats from 96-deep well blocks to 2-L flasks and bioreactors
LV-MAC Production Medium is ready to use, with no supplementation required. The chemically defined formulation results in high reproducibility and lot-to-lot reliability. The medium is not recommended for adherent cell cultures.

Specifications
Viral Production Cell-Line | |
Liquid | |
LV-MAX™ | |
Room Temperature | |
Store at 2–8°C, protect from light. |
Animal Origin-free, Chemically-defined, Protein-free, Serum-free | |
Lentiviral Production Medium | |
1 L | |
No antibiotics | |
Suspension Cell Culture |
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
2616954 | Certificate of Analysis | 29/01/2025 | A3583401 |
2812916 | Certificate of Analysis | 28/01/2025 | A3583401 |
2822826 | Certificate of Analysis | 28/01/2025 | A3583401 |
2871819 | Certificate of Analysis | 26/01/2025 | A3583401 |
2939953 | Certificate of Analysis | 25/01/2025 | A3583401 |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only. Not for use in diagnostic procedures.