missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ Medium 199, no phenol red
GREENER_CHOICE

Medium 199 was originally developed for nutritional studies of chick embryo fibroblasts. It has broad species applicability, particularly for cultivation of non-transformed cells.

Brand:  Gibco™ 11043023

Product Code. 11510386

  • 78.50 € / 500mL

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

Medium 199 was originally developed for nutritional studies of chick embryo fibroblasts. It has broad species applicability, particularly for cultivation of non-transformed cells. Medium 199 is widely used in virology, vaccine production, and in vitro cultivation of primary explants of mouse pancreatic epithelium, and rat lens tissues. We offer a variety of Gibco™ Medium 199 modifications for a range of cell culture applications. Find the right formulation using the media selector tool.

This M199 is modified as follows:
With: L-glutamine
Without: Phenol Red, HEPES

The complete formulation is available.

Compared to other basal media, Medium 199 contains unique components, including adenine, adenosine, hypoxanthine, thymine, and additional vitamins. Medium 199 is available with Earle's salts for use in a CO2 incubator, or with Hanks' salts for use without CO2.

Product Use
For Research Use Only: Not intended for animal or human diagnostic or therapeutic use.

Medium 199 contains no proteins, lipids, or growth factors. Medium 199 uses a sodium bicarbonate buffer system (2.2 g/L) and therefore requires a 5–10% CO2 environment to maintain physiological pH.

Order Info

Shipping Condition: Room Temperature

Compliance

Gibco Medium 199 is manufactured at a cGMP compliant facility located in Grand Island, New York. The facility is registered with the FDA as a medical device manufacturer and is certified to ISO 13485 standards.

TRUSTED_SUSTAINABILITY
Specifications

Specifications

Storage conditions: 2-8° C. Protect from light.
Shipping conditions: Ambient.
Shelf life: 12 months from date of manufacture.
Rat epithelial
Animal Origin
Liquid
Sterile-filtered
No Phenol Red, No Sodium Pyruvate, No HEPES
cGMP-compliant under the ISO 13485 standard
Gibco™
Room Temperature
CO2
Chick Embryo Fibroblasts, Mouse Pancreatic Epithelium, Rat Lens Tissues
1 X
Medium 199
Glutamine, Sodium Bicarbonate
Sustainable packaging
270 - 310 mOsm/kg
500 mL
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

For Research Use Only: Not intended for animal or human diagnostic or therapeutic use.