missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Gibco™ Newborn Calf Serum, heat inactivated, New Zealand origin
Click to view available options
Quantity:
100 mL
500 mL
Unit Size:
100mL
500mL
Beschreibung
Gibco™ sera offers excellent value for basic cell culture, specialty research, and specific assays, earning the trust of researchers with consistent quality and award-winning support that helps meet your research needs and budget requirements.
- Sera Category: Secure
- Origin: New Zealand
- Collected from calves 14 days old or less
Cell Culture, Mammalian Cell Culture
Bestellinformation
Shipping Condition: Dry Ice
Spezifikation
Spezifikation
| Content And Storage | Storage conditions: ≤-10°C Shipping conditions: Frozen |
| Species | Cattle/Bovine |
| Age | Newborn (14 days or less) |
| Country of Origin | New Zealand |
| Form | Liquid |
| Format | Bottle |
| Product Type | Newborn Calf Bovine Serum |
| Quantity | 100 mL |
| Serum Treatment | Heat-inactivated |
| Product Line | Gibco |
| Mehr anzeigen |
For research use or further manufacturing use only. Serum and blood proteins are not for direct administration into humans or animals.
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur