missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ Penicillin-Streptomycin (10,000 U/mL)

For prevention of bacterial contamination of cell cultures due to their effective combined action against gram-positive and gram-negative bacteria

Brand:  Gibco™ 15140148


 View more versions of this product

Product Code. 11568876

  • 19.19 € / 20mL

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

Penicillin was originally purified from fungi Penicillium that acts by interfering directly with turnover of bacterial cell wall and indirectly by triggering release of enzymes that alter the cell wall. Streptomycin was originally purified from Streptomyces griseus. It acts by binding to 30S subunit of bacterial ribosome, leading to inhibition of protein synthesis and death in susceptible bacteria.

  • Contains 10,000 units/mL of penicillin
  • 10,000μg/mL of streptomycin in a 10 mM citrate buffer (for pH stability)
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Storage conditions: -5 to -20°C
Shipping conditions: Frozen
Shelf life: 12 months from date of manufacture
Liquid
Sterile-filtered
Prevention of Cell Culture Contamination
Dry Ice
100 X
Antibiotic
Sterile-filtered
20 mL
Product Suggestions

Product Suggestions

Videos
Safety and Handling
Show More
SDS
Documents

Documents

Certificates

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number2585651 Certificate TypeCertificate of Analysis Date31/01/2025 Catalog Number15140148,15140122
Lot Number243335 Certificate TypeCertificate of Analysis Date31/01/2025 Catalog Number15140148,15140122
Lot Number203043 Certificate TypeCertificate of Analysis Date29/01/2025 Catalog Number15140148,15140122
Lot Number2585619 Certificate TypeCertificate of Analysis Date29/01/2025 Catalog Number15140148,15140122
Lot Number2585609 Certificate TypeCertificate of Analysis Date29/01/2025 Catalog Number15140148,15140122
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Gibco™ Penicillin-Streptomycin (10,000 U/mL) > Quantity: 20 mL

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only. Not for use in diagnostic procedures.