missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ RPMI 1640 Medium, no glutamine
GREENER_CHOICE

RPMI 1640 Medium was originally developed to culture human leukemic cells in suspension and as a monolayer.

33.63 € - 58.25 €

Specifications

Content And Storage Storage conditions: 2-8° C. Protect from light
Shipping conditions: Ambient
Shelf life: 12 months from date of manufacture
Cell Line HeLa, Jurkat, MCF-7, PC-12, PBMC, astrocytes, and carcinomas
Cell Type Leukemic Cells
Classification Animal Origin-free
Concentration 1 X
View More Specs

Requires: Supplementation


Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11340892
GREENER_CHOICE
View Documents
Gibco™
21870076
500 mL
33.63 €
500mL
Please to purchase this item. Need a web account? Register with us today!
11520586
GREENER_CHOICE
View Documents
Gibco™
21870084
1000 mL
58.25 €
1000mL
Please to purchase this item. Need a web account? Register with us today!
Description

Description

RPMI 1640 Medium was originally developed to culture human leukemic cells in suspension and as a monolayer. Roswell Park Memorial Institute (RPMI) 1640 Medium has since been found suitable for a variety of mammalian cells, including HeLa, Jurkat, MCF-7, PC12, PBMC, astrocytes, and carcinomas. We offer a variety of RPMI 1640 Medium modifications for a range of cell culture applications. Find the right formulation using the media selector tool.

This RPMI is modified as follows:

With: Phenol Red

Without: HEPES, L-glutamine

The complete formulation is available.

Using RPMI

RPMI 1640 Medium is unique from other media because it contains the reducing agent glutathione and high concentrations of vitamins. RPMI 1640 Medium contains biotin, vitamin B12, and PABA, which are not found in Eagle's Minimal Essential Medium or Dulbecco's Modified Eagle Medium. In addition, the vitamins inositol and choline are present in very high concentrations. RPMI 1640 Medium contains no proteins, lipids, or growth factors. Therefore, RPMI 1640 Medium requires supplementation, commonly with 10% Fetal Bovine Serum (FBS). RPMI 1640 Medium uses a sodium bicarbonate buffer system (2.0 g/L), and therefore requires a 5–10% CO2 environment to maintain physiological pH.

Order Info

Shipping Condition: Room Temperature

Specifications

Specifications

Storage conditions: 2-8° C. Protect from light
Shipping conditions: Ambient
Shelf life: 12 months from date of manufacture
Leukemic Cells
1 X
RPMI 1640 Medium (Roswell Park Memorial Institute 1640 Medium)
Sterile-filtered
Phenol Red
cGMP-compliant under the ISO 13485 standard
Room Temperature
HeLa, Jurkat, MCF-7, PC-12, PBMC, astrocytes, and carcinomas
Animal Origin-free
Liquid
Standard Serum Supplementation
Sterile-filtered
No Glutamine, No HEPES, No Sodium Pyruvate
Gibco™
Videos
Safety and Handling

Safety and Handling

missing translation for 'shelfLife' : 12 Months From Date of Manufacture

SDS
Documents

Documents

Product Certifications

For Research Use or Further Manufacturing. Not for diagnostic use or direct administration into humans or animals.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.