missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ Schneider's Drosophila Medium
GREENER_CHOICE

Modified to contain L-glutamine and sodium bicarbonate

Brand:  Gibco™ 21720001

764.28 EUR valid until 2024-11-30
Use promo code "21651" to get your promotional price.


 View more versions of this product

Product Code. 15372382

  • 1158.00 € / Pack of 10

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

Schneider's Drosophila Medium was originally designed for the growth of S2 cells from the fruit fly, Drosophila melanogaser. Schneider's Drosophila Medium can also be used to culture other dipteran cell lines. This medium can be used for transient or stable transfection and large scale protein production in S2 cells.

Formulation

  • Contains unique components required for S2 cell culture, including malic acid, fumaric acid, alpha-ketoglutaric acid, succinic acid, and the sugar trehelos
  • Commonly supplemented with 10% fetal bovine serum (FBS) before use
  • Requires ambient CO2 in order to maintain physiological pH

Cell Culture, Drosophila S2 Cell Culture, Insect Cell Culture

Compliance

Gibco Schneider's Drosophila Medium is manufactured at a cGMP-compliant facility located in Paisley, Scotland, UK. The facility is registered with the FDA as a medical device manufacturer and is certified to the ISO 13485 standard.

TRUSTED_SUSTAINABILITY
Specifications

Specifications

Storage conditions: 2-8 °C. Protect from light
Shipping conditions: Ambient
Shelf life: 12 months from date of manufacture
Insect Cell
Schneider's Drosophila Medium
Glutamine
10 x 500 mL
D.Mel-2
Liquid
Sterile
No HEPES, No Phenol Red, No Sodium Pyruvate
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

For Research Use Only. Not for use in diagnostic procedures.