missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ Schneider's Drosophila Medium
GREENER_CHOICE

Product Code. p-4924981
Click to view available options
Quantity:
500 mL
10 x 500 mL
Unit Size:
500mL
Pack of 10
This item is not returnable. View return policy

Product Code. 11590576

Brand: Gibco™ 21720024

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Modified to contain L-glutamine and sodium bicarbonate

Schneider's Drosophila Medium was originally designed for the growth of S2 cells from the fruit fly, Drosophila melanogaser. Schneider's Drosophila Medium can also be used to culture other dipteran cell lines. This medium can be used for transient or stable transfection and large scale protein production in S2 cells.

Formulation

  • Contains unique components required for S2 cell culture, including malic acid, fumaric acid, alpha-ketoglutaric acid, succinic acid, and the sugar trehelos
  • Commonly supplemented with 10% fetal bovine serum (FBS) before use
  • Requires ambient CO2 in order to maintain physiological pH

Cell Culture, Drosophila S2 Cell Culture, Insect Cell Culture

Compliance

Gibco Schneider's Drosophila Medium is manufactured at a cGMP-compliant facility located in Paisley, Scotland, UK. The facility is registered with the FDA as a medical device manufacturer and is certified to the ISO 13485 standard.

TRUSTED_SUSTAINABILITY

Specifications

Cell Line S2 (Schneider 2)
Cell Type Insect Cell
Form Liquid
Product Type Schneider's Drosophila Medium
With Additives Glutamine
Without Additives No HEPES, No Phenol Red, No Sodium Pyruvate
Product Line Gibco
Quantity 500 mL
Shipping Condition Room Temperature
Content And Storage Storage conditions: 2-8°C. Protect from light
Shipping conditions: Ambient
Shelf life: 12 months from date of manufacture

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.

Vielen Dank, dass Sie uns helfen, unsere Website zu verbessern. Ihr Feedback wurde übermittelt