missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Gibco™ Sf-900™ III SFM
A low-hydrolysate, serum-free, protein-free, animal origin–free insect cell culture medium
Brand: Gibco™ 12658019
68.60 EUR valid until 2024-11-30
Use promo code "21651" to get your promotional price.
This item is not returnable.
View return policy
Description
- Sf-900™ III SFM is a low-hydrolysate, serum-free, protein-free, animal origin–free insect cell culture medium optimized for the growth and maintenance of Spodoptera frugiperda (Sf9 and Sf21) cells
- Also optimized for recombinant gene expression using the baculovirus and stable insect expression systems
- Medium is suitable for suspension and monolayer culture methods and supports growth of other lepidopteran cell lines
- Spodoptera frugiperda (Sf9) cells grown in Sf-900™ III SFM achieve maximum cell densities of 10 to 14 × 106 cells/mL
- Sf-900™ III SFM is an improved serum-free, protein-free, animal origin–free medium designed for growth of Sf9 and other lepidopteran cell lines and production of insect viruses and recombinant proteins
- Ready to use format does not require addition of serum, glutamine, or surfactants
- Cells adapted to other commercially available serum-free media should be sequentially adapted into Sf-900™ III SFM=
- Reduced hydrolysate concentrations improve lot-to-lot consistency
- Store at 2°C to 8°C, protected from light
Order Info
Shipping Condition: Room temperature
Compliance
Gibco Sf-900 II SFM is manufactured at a cGMP-compliant facility located in Grand Island, New York. The facility is registered with the FDA as a medical device manufacturer and is certified to ISO 13485 standards.
Specifications
Storage conditions: 2–8°C. Protect from light Shipping conditions: Ambient Shelf life: 18 months from date of manufacture |
|
Insect Cell | |
Liquid | |
Serum-free | |
Sustainable packaging | |
500 mL | |
S. frugiperda, Spodoptera frugiperda |
Sf21, Sf9 | |
Animal Origin-free, Protein-free, Serum-free | |
Insect Cell Serum Free Medium (SFM) | |
Glutamine | |
Gibco™, Sf-900™ | |
Room Temperature |
For Research Use Only. Not for use in diagnostic procedures.