missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ Sf-900™ III SFM

A low-hydrolysate, serum-free, protein-free, animal origin–free insect cell culture medium

Brand:  Gibco™ 12658001

1261.40 EUR valid until 2024-11-30
Use promo code "21651" to get your promotional price.


 View more versions of this product

Product Code. 15236025

  • 1802.00 € / 20 L

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

  • Sf-900™ III SFM is a low-hydrolysate, serum-free, protein-free, animal origin–free insect cell culture medium optimized for the growth and maintenance of Spodoptera frugiperda (Sf9 and Sf21) cells
  • Also optimized for recombinant gene expression using the baculovirus and stable insect expression systems
  • Medium is suitable for suspension and monolayer culture methods and supports growth of other lepidopteran cell lines
  • Spodoptera frugiperda (Sf9) cells grown in Sf-900™ III SFM achieve maximum cell densities of 10 to 14 × 106 cells/mL
  • Sf-900™ III SFM is an improved serum-free, protein-free, animal origin–free medium designed for growth of Sf9 and other lepidopteran cell lines and production of insect viruses and recombinant proteins
  • Ready to use format does not require addition of serum, glutamine, or surfactants
  • Cells adapted to other commercially available serum-free media should be sequentially adapted into Sf-900™ III SFM=
  • Reduced hydrolysate concentrations improve lot-to-lot consistency
  • Store at 2°C to 8°C, protected from light

Order Info

Shipping Condition: Room temperature

Compliance

Gibco Sf-900 II SFM is manufactured at a cGMP-compliant facility located in Grand Island, New York. The facility is registered with the FDA as a medical device manufacturer and is certified to ISO 13485 standards.

TRUSTED_SUSTAINABILITY
Specifications

Specifications

Storage conditions: 2–8°C. Protect from light
Shipping conditions: Ambient
Shelf life: 18 months from date of manufacture
Sf21, Sf9
Animal Origin-free
Suspension Cell Culture
Insect Cell Serum Free Medium (SFM)
Sterile
No Phenol Red, No Sodium Pyruvate, No Sodium Bicarbonate
Gibco™, Sf-900™
20 L
Non-CO2
Insect Cell
1 X
Liquid
Serum-free
Glutamine
Low
Cell Culture Grade
Room Temperature
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

For Research Use Only. Not for use in diagnostic procedures.