Learn More
Gibco™ Sf-900™ Medium (1.3X)

Description
Sf-900 SFM is developed for the convenient and reproducible formation of 1% agarose overlays used for plaque assays of baculovirus in Spodoptera frugiperda (Sf9 and Sf21) cells. When used with Gibco 4% Agarose Gel, Sf900™ SFM (1.3x) allows for convenient formation of a 1% overlay with correct 1x final osmolality and nutrition required for insect cell growth during the plaque assay.
Baculovirus Production and Titering, Bioproduction, Cell Culture, Insect Cell Culture, Insect Expression, Protein Expression, Proteins, Expression, Isolation and Analysis, Sf9 and Sf21 Cell Culture, Viral Vaccine Production
Order Info
Shipping Condition: Room temperature
Compliance
Gibco Sf-900 II SFM is manufactured at a cGMP compliant facility located in Grand Island, New York. The facility is registered with the FDA as a medical device manufacturer and is certified to ISO 13485 standards.
Specifications
Specifications
| Cell Line | Sf21, Sf9 |
| Cell Type | Insect Cell |
| Form | Liquid |
| Product Type | Insect Cell Serum Free Medium (SFM) |
| With Additives | Glutamine |
| Product Line | Gibco, Sf-900 |
| Quantity | 100 mL |
| Shipping Condition | Room Temperature |
| Species | S. frugiperda, Spodoptera frugiperda |
| Content And Storage | Storage conditions: 2-8°C. Protect from light Shipping conditions: Ambient Shelf life: 9 months from date of manufacture |
For Research Use Only. Not for use in diagnostic procedures.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.