missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ Sf9 cells in Sf-900™ II SFM

Used to isolate and propagate recombinant baculoviral stocks and to produce recombinant proteins

Brand:  Gibco™ 11496015

Product Code. 10500343

  • 838.00 € / Each

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers

Alert:

This product contains Dimethyl Sulfoxide (DMSO), a hazardous material. Review the Material Safety Data Sheet before handling.

This item is not returnable. View return policy

Description

Description

Gibco Sf9 cells are adapted to serum-free suspension culture in Gibco Sf-900 II SFM.

  • Recombinant protein expression from a variety of expression systems
  • Good growth in adherent or suspension culture
  • Small, regular size that generates even monolayers and plaques
  • Documented lineage from a low passage Master Cell Bank
  • Quality and performance testing

Baculovirus Production and Titering, Bioproduction, Cell Culture, Insect Cell Culture, Insect Expression, Insect Expression Cell Lines, Protein Expression, Proteins, Expression, Isolation and Analysis, Sf9 and Sf21 Cell Culture, Viral Vaccine Production

Order Info

Shipping Condition: Dry Ice

TRUSTED_SUSTAINABILITY
Specifications

Specifications

1 x 1.5 mL (1.0 x 107 cells/mL)

Storage conditions: Liquid nitrogen (vapor phase)

Insect Cells
S. frugiperda
Insect Cells
Dry Ice
Sf9
Liquid
Sf-900 II SFM (Serum-Free Media)
1.5 mL
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only. Not for use in diagnostic procedures.