missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ Distilled Water
GREENER_CHOICE

High-quality water for use as a solvent in the preparation of cell culture media and laboratory reagents

30.89 € - 161.00 €

Specifications

Grade Cell Culture
Packaging Bottle
Sterility Sterile-filtered
Treatment(s) Not DEPC-Treated
Purification Method Membrane-Filtered
View More Specs

Products 4
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
12589069
GREENER_CHOICE
View Documents
Gibco™
15230162
500 mL
30.89 €
500mL
Please to purchase this item. Need a web account? Register with us today!
15697708
GREENER_CHOICE
View Documents
Gibco™
15230147
1000 mL
65.00 €
1000mL
Please to purchase this item. Need a web account? Register with us today!
15480594
GREENER_CHOICE
View Documents
Gibco™
15230204
10 x 500 mL
161.00 €
Pack of 10
Please to purchase this item. Need a web account? Register with us today!
15303298
GREENER_CHOICE
View Documents
Gibco™
15230001
6 x 1 L
137.00 €
Each
Please to purchase this item. Need a web account? Register with us today!
Description
Show More
Specifications

Specifications

Cell Culture
Sterile-filtered
Membrane-Filtered
Bottle
Not DEPC-Treated
Room Temperature
Videos
Safety and Handling

Safety and Handling

missing translation for 'shelfLife' : 24 Months

Recommended Storage : Storage conditions: 15 – 30°C
Shipping conditions: Ambient
Shelf life: 24 months from date of manufacture

SDS
Documents

Documents

Product Certifications

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Gibco™ Distilled Water

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.