Learn More
Gibco™ TrypLE™ Express Enzyme (1X), no phenol red
Unlike porcine trypsin, free of animal-derived components
Brand: Gibco™ 12604054
682.10 EUR valid until 2025-02-21
Use promo code "24115" to get your promotional price.
Description
Cleaves peptide bonds on the C-terminal sides of lysine and arginine, and is a direct replacement for trypsin. Exceptional purity increases specificity and reduces damage to cells that can be caused by other enzymes present in some trypsin extracts. Used for dissociating a wide range of adherent mammalian cells, including CHO, HEK 293, A529, primary human keratinocytes, and embryonic stem cells.
- Gentle on cells
- Room temperature-stable for 24 months
- Easy to use
- Animal origin-free
- Gentle on cells, purity increases specificity due to the action of a single enzyme and reduces damage caused by cleavage from multiple enzymes in trypsin and other extracts
- Can be directly substituted for trypsin in existing protocols
- Dilution alone inactivates TrypLE Express, avoiding the need for trypsin inhibitors, such as FBS
Energy efficient, less hazardous, sustainable packaging
Cell Culture, Mammalian Cell Culture
Order Info
Shipping Condition: Room Temperature
Compliance
Manufactured at a cGMP-compliant facility that is registered with the FDA as a medical device manufacturer and is certified to ISO 13485 standards.
Specifications
EDTA | |
Mammalian Cells | |
1 X | |
Liquid | |
Sterile-filtered | |
Energy efficient, Sustainable packaging | |
Gibco™ | |
Room Temperature Stable | |
Animal origin-free |
Storage conditions: 15°C to 30°C, protect from light Alternate storage conditions: 2°C to 8°C or -5 to -20°C Shipping conditions: Room temperature Shelf life: 24 months from date of manufacture |
|
Animal Origin-free | |
Adherent Cell Culture | |
Cell Culture Dissociation Reagent | |
No Phenol Red | |
Check and Record | |
5 L | |
Room Temperature | |
In vitro Bioassay |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only. Not for use in diagnostic procedures.