missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ TrypLE™ Express Enzyme (1X), phenol red
GREENER_CHOICE

Animal origin-free, recombinant enzyme

32.90 € - 556.00 €

Specifications

Chelators EDTA
Cell Type Mammalian Cells
Classification Animal Origin-free
Concentration 1 X
Culture Type Adherent Cell Culture
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
10043382
GREENER_CHOICE
View Documents
Gibco™
12605010
100 mL
32.90 €
100mL
Please to purchase this item. Need a web account? Register with us today!
11568856
GREENER_CHOICE
View Documents
Gibco™
12605036
20 x 100mL
556.00 €
Pack of 20
Please to purchase this item. Need a web account? Register with us today!
11558856
GREENER_CHOICE
View Documents
Gibco™
12605028
500 mL
130.00 €
500mL
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Used for dissociating a wide range of adherent mammalian cells, including CHO, HEK 293, A529, primary human keratinocytes, and embryonic stem cells. TrypLE™ Express cleaves peptide bonds on the C-terminal sides of lysine and arginine, and is a direct replacement for trypsin. Exceptional purity increases specificity and reduces damage to cells that can be caused by other enzymes present in some trypsin extracts.

  • Gentle On Cells
  • urity increases specificity due to the action of single enzyme, reducing damage caused by cleavage from enzymes present in some trypsin preparations and in other protease extracts
  • Room-Temperature Stable for 24 months
  • Ready to use
  • Can be directly substituted for trypsin in existing protocols
  • Dilution alone inactivates TrypLE Express, avoiding the need for trypsin inhibitors, such as FBS

Energy efficient, less hazardous, sustainable packaging

Cell Culture, Mammalian Cell Culture

Order Info

Shipping Condition: Room Temperature

Compliance

Manufactured in facilities compliant with cGMP manufacturing requirements, certified to the ISO 13485 standard, and registered with the FDA as medical device manufacturers.

Specifications

Specifications

EDTA
Animal Origin-free
Adherent Cell Culture
Cell Culture Dissociation Reagent
Phenol Red, EDTA
Check and Record
Room Temperature Stable
Animal origin-free
Mammalian Cells
1 X
Liquid
Sterile-filtered
Energy efficient, Sustainable packaging
Gibco™
Room Temperature
In vitro Bioassay
Videos
Safety and Handling

Safety and Handling

missing translation for 'shelfLife' : 24 Months

Product Identifier
  • 1-(3-Methoxyphenyl)piperazine dihydrochloride

Signal Word
  • Warning

Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3

Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.

Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

Supplemental information
  • MIXTURE LIST-Contains : N-(3-Methoxyphenyl)piperazine

SDS
Documents

Documents

Product Certifications

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.